Order Entry
Austria
ContactUsLinkComponent
Clomipramine Related Compounds, Reference Standard
Clomipramine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Clomipramine Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    2305824-51-3
  • Synonym(s):
    N1-[3-(3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)propyl]-N1,N3,N3-trimethylpropane-1,3-diamine dihydrochloride
  • Description:
    Clomipramine Related Compound A
  • Formula:
    C₁₉H₂₃Cl₃N₂

Specifications

Frequently Bought Together