Order Entry
Austria
Orders LinkContactUsLinkComponent
Clindamycin Phosphate, Reference Standards

Clindamycin Phosphate, Reference Standards

Clindamycin Phosphate, USP reference standard, Vial, 250 mg
Catalog # 1138008.
Supplier:   USP
Clindamycin Phosphate, Reference Standards
Catalog # 1138008.
Supplier:  USP
CAS Number:  
Clindamycin Phosphate, Reference Standards

Clindamycin Phosphate, USP reference standard, Vial, 250 mg

CAS Number:  
Price
537,00 €
Log in for availability
1 * 250 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    250 mg
  • Standards:
    USP reference standard
  • Description:
    Clindamycin Phosphate
  • Cat. No.:
    1138008.
  • Formula:
    C₁₈H₃₄ClN₂O₈PS
  • Description:
    Clindamycin Phosphate
  • CAS No.:
    24729-96-2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.