Order Entry
Austria
ContactUsLinkComponent
Acetone, CHROMASOLV™ E for pesticide residue analysis, Riedel-de Haën™
Irritant
Flammable
Acetone, CHROMASOLV™ E for pesticide residue analysis, Riedel-de Haën™
Certificates
Irritant
Flammable
Acetone, CHROMASOLV™ E for pesticide residue analysis, Riedel-de Haën™

Specification Test Results

  • Assay (GC):
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Suitability for GC-MS of dioxines, furanes, PCPs:
    complying

Specification Test Results

Frequently Bought Together