Order Entry
Austria
Orders LinkContactUsLinkComponent
Zolmitriptan, Reference Standards

Zolmitriptan, Reference Standards

Anbieter:   USP
Zolmitriptan, Reference Standards
Anbieter:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1727009.
  • 1727018.
  • Description:
  • Zolmitriptan
  • Zolmitriptan R-Isomer
  • Synonym(s):
  • (R)-4-({3-[2-(Dimethylamino)ethyl]indol-5-yl}methyl)oxazolidin-2-one
  • CAS No.:
  • 139264-17-8
  • 139264-24-7

Specifications

Product Family Options

Product Information

  • Description
    Standards
    Pack type
    Pk
    Verfügbarkeit
    Preis
  • Zolmitriptan
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 399,00 €
    399,00 €
    1 * 200 mg
       399,00 €

    Specifications:
    Mehr
    Specifications
    Cat. No.
    1727009.
    Description
    Zolmitriptan
    Synonym(s)
    CAS No.
    139264-17-8
  • Zolmitriptan R-Isomer
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 290,00 €
    1 290,00 €
    1 * 20 mg
       1 290,00 €

    Specifications:
    Mehr
    Specifications
    Cat. No.
    1727018.
    Description
    Zolmitriptan R-Isomer
    Synonym(s)
    (R)-4-({3-[2-(Dimethylamino)ethyl]indol-5-yl}methyl)oxazolidin-2-one
    CAS No.
    139264-24-7
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.