Order Entry
Austria
Orders LinkContactUsLinkComponent
Ubidecarenone, Reference Standards

Ubidecarenone, Reference Standards

Anbieter:   USP
Ubidecarenone, Reference Standards
Anbieter:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Caution: Cold Shipment required

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1705301.
  • 1705312.
  • 1705323.
  • Description:
  • Ubidecarenone
  • Ubidecarenone for System Suitability
  • Ubidecarenone Related Compound A
  • Synonym(s):
  • Coenzyme Q9
  • CAS No.:
  • 303-98-0
  • 303-98-0
  • 303-97-9

Specifications

Product Family Options

Product Information

  • Description
    Standards
    Pack type
    Pk
    Verfügbarkeit
    Preis
  • Ubidecarenone
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 560,00 €
    560,00 €
    1 * 200 mg
       560,00 €

    Specifications:
    Mehr
    Specifications
    Cat. No.
    1705301.
    Description
    Ubidecarenone
    Synonym(s)
    CAS No.
    303-98-0
  • Ubidecarenone for System Suitability
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 060,00 €
    1 060,00 €
    1 * 25 mg
       1 060,00 €

    Specifications:
    Mehr
    Specifications
    Cat. No.
    1705312.
    Description
    Ubidecarenone for System Suitability
    Synonym(s)
    CAS No.
    303-98-0
  • Ubidecarenone Related Compound A
    USP reference standard
    Ampule
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 070,00 €
    1 070,00 €
    1 * 15 mg
       1 070,00 €

    Specifications:
    Mehr
    Specifications
    Cat. No.
    1705323.
    Description
    Ubidecarenone Related Compound A
    Synonym(s)
    Coenzyme Q9
    CAS No.
    303-97-9