Order Entry
Austria
Orders LinkContactUsLinkComponent
Polyvinyl Acetate, Reference Standards

Polyvinyl Acetate, Reference Standards

Supplier:   USP
Polyvinyl Acetate, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1548032.
  • 1548043.
  • Description:
  • Polyvinyl Acetate
  • Polyvinyl Acetate Dispersion
  • Synonym(s):
  • Polyvinyl Acetate stabilized with Povidone and Sodium Lauryl Sulfate
  • CAS No.:
  • 9003-20-7
  • 9003-20-7

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1548032.
    Polyvinyl Acetate
    Polyvinyl Acetate
    USP reference standard
    Vial
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 331,00 €
    331,00 €
    1 * 1 g
       331,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1548032.
    Description
    Polyvinyl Acetate
    Synonym(s)
    CAS No.
    9003-20-7
  • 1548043.
    Polyvinyl Acetate Dispersion
    Polyvinyl Acetate Dispersion
    USP reference standard
    Ampule
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 411,00 €
    411,00 €
    1 * 1 g
       411,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1548043.
    Description
    Polyvinyl Acetate Dispersion
    Synonym(s)
    Polyvinyl Acetate stabilized with Povidone and Sodium Lauryl Sulfate
    CAS No.
    9003-20-7