Order Entry
Austria
ContactUsLinkComponent
Pergolide Related Compounds, Reference Standard
Pergolide Related Compounds, Reference Standard
Supplier:  USP
Certificates
Pergolide Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    66104-23-2
  • UN No.:
    1544
  • Description:
    Pergolide Mesylate

Specifications

Frequently Bought Together