Methoxsalen, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1417001.
- 1065345.
- 1349740.
- Description:
- Methoxsalen
- Methoxsalen Related Compound A
- Methoxsalen Related Compound B
- Synonym(s):
- 4-Methoxy-7H-furo[3,2-g]chromen-7-one
- 4,9-Dimethoxy-7H-furo[3,2-g]chromen-7-one
- CAS No.:
- 298-81-7
- 484-20-8
- 482-27-9
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1417001.MethoxsalenSpecifications:
More
Specifications
Cat. No.1417001.DescriptionMethoxsalenSynonym(s)CAS No.298-81-7 - 1065345.Methoxsalen Related Compound AMethoxsalen Related Compound AUSP reference standardVial25 mgLog in for availability1 * 25 mg1 220,00 €Specifications:
More
Specifications
Cat. No.1065345.DescriptionMethoxsalen Related Compound ASynonym(s)4-Methoxy-7H-furo[3,2-g]chromen-7-oneCAS No.484-20-8 - 1349740.Methoxsalen Related Compound BMethoxsalen Related Compound BUSP reference standardVial25 mgLog in for availability1 * 25 mg1 130,00 €Specifications:
More
Specifications
Cat. No.1349740.DescriptionMethoxsalen Related Compound BSynonym(s)4,9-Dimethoxy-7H-furo[3,2-g]chromen-7-oneCAS No.482-27-9
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.



