Order Entry
Austria
Orders LinkContactUsLinkComponent
Lactic Acid, Reference Standard

Lactic Acid, Reference Standard

Lactic Acid, USP reference standard, 1.5 mL/ampule; 3 ampules, Ampule, 3
Artikelnr. 1356734.
Anbieter:   USP
Lactic Acid, Reference Standard
Artikelnr. 1356734.
Anbieter:  USP
CAS Nummer:  
Lactic Acid, Reference Standard

Lactic Acid, USP reference standard, 1.5 mL/ampule; 3 ampules, Ampule, 3

CAS Nummer:  
Preis
441,00 €
Log in for availability
1 * 3 ST
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications

Specifications

  • Pack type:
    Ampule
  • Pk:
    3
  • Standards:
    USP reference standard
  • Description:
    Lactic Acid
  • Pk Info:
    1.5 mL/ampule; 3 ampules
  • UN No.:
    3265
  • Cat. No.:
    1356734.
  • Description:
    Lactic Acid
  • CAS No.:
    50-21-5

Specifications