Order Entry
Austria
Orders LinkContactUsLinkComponent
Isoflurane, Reference Standards

Isoflurane, Reference Standards

Supplier:   USP
Isoflurane, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1349014.
  • 1349025.
  • 1349003.
  • Description:
  • Isoflurane Related Compound A
  • Isoflurane Related Compound B
  • Isoflurane
  • Synonym(s):
  • 1-Chloro-2,2,2-trifluoroethyl chlorodifluoromethyl ether
  • 2,2,2-Trifluoroethyl difluoromethyl ether
  • CAS No.:
  • 32778-08-8
  • 1885-48-9
  • 26675-46-7
  • Formula:
  • C₃HCl₂F₅O
  • C₃H₃F₅O
  • UN No.:
  • 3334
  • 3334
  • 3334

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1349014.
    Isoflurane Related Compound A
    Isoflurane Related Compound A
    USP reference standard
    Ampule
    0,1 mL
    Log in for availability
    Log in for availability
    1 * 0,1 mL
    1 * 0,1 mL - 1 360,00 €
    1 360,00 €
    1 * 0,1 mL
       1 360,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1349014.
    Description
    Isoflurane Related Compound A
    Synonym(s)
    1-Chloro-2,2,2-trifluoroethyl chlorodifluoromethyl ether
    CAS No.
    32778-08-8
    Formula
    C₃HCl₂F₅O
    UN No.
    3334
  • 1349025.
    Isoflurane Related Compound B
    Isoflurane Related Compound B
    USP reference standard
    Ampule
    0,1 mL
    Log in for availability
    Log in for availability
    1 * 0,1 mL
    1 * 0,1 mL - 1 360,00 €
    1 360,00 €
    1 * 0,1 mL
       1 360,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1349025.
    Description
    Isoflurane Related Compound B
    Synonym(s)
    2,2,2-Trifluoroethyl difluoromethyl ether
    CAS No.
    1885-48-9
    Formula
    C₃H₃F₅O
    UN No.
    3334
  • 1349003.
    Isoflurane
    Isoflurane
    USP reference standard
    Ampule
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 354,00 €
    354,00 €
    1 * 1 mL
       354,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1349003.
    Description
    Isoflurane
    Synonym(s)
    CAS No.
    26675-46-7
    Formula
    UN No.
    3334
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.