Order Entry
Austria
Orders LinkContactUsLinkComponent
Polyethylene, Reference Standards

Polyethylene, Reference Standards

Supplier:   USP
Polyethylene, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Zertifizierung: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1546809.
  • 1546707.
  • Description:
  • Low-Density Polyethylene
  • High-Density Polyethylene
  • CAS No.:
  • 9002-88-4
  • 9002-88-4
  • Formula:
  • C₂H₄
  • C₂H₄

Specifications

Product Family Options

Product Information

  • Standards
    Pk Info
    Pack type
    Pk
    Availability
    Price
  • 1546809.
    Low-Density Polyethylene
    Low-Density Polyethylene
    USP reference standard
    3 strips
    Strips
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 448,00 €
    448,00 €
    1 * 1 KIT
       448,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1546809.
    Description
    Low-Density Polyethylene
    CAS No.
    9002-88-4
    Formula
    C₂H₄
  • 1546707.
    High-Density Polyethylene
    High-Density Polyethylene
    USP reference standard
    3 strips 2'' x 2 1/2'' each
    Strips
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 412,00 €
    412,00 €
    1 * 1 KIT
       412,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1546707.
    Description
    High-Density Polyethylene
    CAS No.
    9002-88-4
    Formula
    C₂H₄