Order Entry
Austria
Orders LinkContactUsLinkComponent
Glipizide, Reference Standards

Glipizide, Reference Standards

Supplier:   USP
Glipizide, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Zertifizierung: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1292507.
  • 1292609.
  • 1292610.
  • 1292631.
  • Description:
  • Glipizide
  • Glipizide Related Compound A
  • Glipizide Related Compound B
  • Glipizide Related Compound C
  • Synonym(s):
  • N-{2-[(4-aminosulfonyl)phenyl]ethyl}-5-methyl-pyrazinecarboxamide
  • 6-methyl-N-[2-(4-sulphamoylphenyl)ethyl]pyrazine-2-carboxamide
  • 1-cyclohexyl-3-[[4-[2-[[(6-methylpyrazin-2-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]urea
  • CAS No.:
  • 29094-61-9
  • 33288-71-0
  • 66375-96-0

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1292507.
    Glipizide
    Glipizide
    USP reference standard
    Vial
    125 mg
    Log in for availability
    Log in for availability
    1 * 125 mg
    1 * 125 mg - 324,00 €
    324,00 €
    1 * 125 mg
       324,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1292507.
    Description
    Glipizide
    Synonym(s)
    CAS No.
    29094-61-9
  • 1292609.
    Glipizide Related Compound A
    Glipizide Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 270,00 €
    1 270,00 €
    1 * 25 mg
       1 270,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1292609.
    Description
    Glipizide Related Compound A
    Synonym(s)
    N-{2-[(4-aminosulfonyl)phenyl]ethyl}-5-methyl-pyrazinecarboxamide
    CAS No.
    33288-71-0
  • 1292610.
    Glipizide Related Compound B
    Glipizide Related Compound B
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 110,00 €
    1 110,00 €
    1 * 15 mg
       1 110,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1292610.
    Description
    Glipizide Related Compound B
    Synonym(s)
    6-methyl-N-[2-(4-sulphamoylphenyl)ethyl]pyrazine-2-carboxamide
    CAS No.
  • 1292631.
    Glipizide Related Compound C
    Glipizide Related Compound C
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 210,00 €
    1 210,00 €
    1 * 15 mg
       1 210,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1292631.
    Description
    Glipizide Related Compound C
    Synonym(s)
    1-cyclohexyl-3-[[4-[2-[[(6-methylpyrazin-2-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]urea
    CAS No.
    66375-96-0
Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.