Order Entry
Austria
Orders LinkContactUsLinkComponent
Elemental Impurities Mixture, Reference Standards | Avantor

Elemental Impurities Mixture, Reference Standards

Anbieter:   USP
Elemental Impurities Mixture, Reference Standards
Anbieter:  USP
Certificates
Elemental Impurities Mixture, Reference Standards

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1234410.
  • 1234427.
  • 1234431.
  • Description:
  • Elemental Impurities Class 1 and 2A Mixture
  • Elemental Impurities Class 2B Mixture
  • Elemental Impurities Class 3 Mixture
  • UN No.:
  • 1789
  • 1789
  • 2031

Specifications

Product Family Options

Product Information

  • Description
    Standards
    Pack type
    Pk
    Verfügbarkeit
    Preis
  • Elemental Impurities Class 1 and 2A Mixture
    USP reference standard
    Bottle
    120 mL
    Log in for availability
    Log in for availability
    1 * 120 mL
    1 * 120 mL - 527,00 €
    527,00 €
    1 * 120 mL
       527,00 €

    Specifications:
    Mehr
    Specifications
    Cat. No.
    1234410.
    Description
    Elemental Impurities Class 1 and 2A Mixture
    UN No.
    1789
  • Elemental Impurities Class 2B Mixture
    USP reference standard
    Bottle
    120 mL
    Log in for availability
    Log in for availability
    1 * 120 mL
    1 * 120 mL - 576,00 €
    576,00 €
    1 * 120 mL
       576,00 €

    Specifications:
    Mehr
    Specifications
    Cat. No.
    1234427.
    Description
    Elemental Impurities Class 2B Mixture
    UN No.
    1789
  • Elemental Impurities Class 3 Mixture
    USP reference standard
    Bottle
    120 mL
    Log in for availability
    Log in for availability
    1 * 120 mL
    1 * 120 mL - 546,00 €
    546,00 €
    1 * 120 mL
       546,00 €

    Specifications:
    Mehr
    Specifications
    Cat. No.
    1234431.
    Description
    Elemental Impurities Class 3 Mixture
    UN No.
    2031
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.