Valacyclovir, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Zertifizierung: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Zertifizierung: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1707839.
- 1707817.
- Description:
- Valacyclovir Hydrochloride
- D-Valacyclovir
- Synonym(s):
- D-Valine, 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy] ethyl ester
- CAS No.:
- 502421-45-6
- 142963-60-8
- Formula:
- C₁₃H₂₁ClN₆O₄
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1707839.Valacyclovir HydrochlorideValacyclovir HydrochlorideUSP reference standardVial200 mgLog in for availability1 * 200 mg412,00 €Specifications:
More
Specifications
Cat. No.1707839.DescriptionValacyclovir HydrochlorideSynonym(s)CAS No.502421-45-6FormulaC₁₃H₂₁ClN₆O₄ - 1707817.D-ValacyclovirSpecifications:
More
Specifications
Cat. No.1707817.DescriptionD-ValacyclovirSynonym(s)D-Valine, 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy] ethyl esterCAS No.142963-60-8Formula
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.



