Order Entry
Austria
Orders LinkContactUsLinkComponent
Docusate Sodium, Reference Standards

Docusate Sodium, Reference Standards

Supplier:   USP
Docusate Sodium, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1224802.
  • 1224824.
  • Description:
  • Docusate Sodium
  • Docusate Sodium Related Compound B
  • Synonym(s):
  • disodium mono (2-ethylhexyl) sulfosuccinate
  • CAS No.:
  • 577-11-7

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • Docusate Sodium
    USP reference standard
    Vial
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 456,00 €
    456,00 €
    1 * 1 g
       456,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1224802.
    Description
    Docusate Sodium
    Synonym(s)
    CAS No.
    577-11-7
  • 1224824.
    Docusate Sodium Related Compound B
    Docusate Sodium Related Compound B
    USP reference standard
    Vial
    40 mg
    Log in for availability
    Log in for availability
    1 * 40 mg
    1 * 40 mg - 1 110,00 €
    1 110,00 €
    1 * 40 mg
       1 110,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1224824.
    Description
    Docusate Sodium Related Compound B
    Synonym(s)
    disodium mono (2-ethylhexyl) sulfosuccinate
    CAS No.