Order Entry
Austria
Orders LinkContactUsLinkComponent
Cumene, Reference Standard

Cumene, Reference Standard

Cumene, USP reference standard, 1.2 mL/ampule; 3 ampules, Ampule, 3 Ampoul
Artikelnr. 1151709.
Anbieter:   USP
Cumene, Reference Standard
Artikelnr. 1151709.
Anbieter:  USP
CAS Nummer:  
Cumene, Reference Standard

Cumene, USP reference standard, 1.2 mL/ampule; 3 ampules, Ampule, 3 Ampoul

CAS Nummer:  
Preis
250,00 €
Log in for availability
1 * 3 Ampull
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Ampule
  • Pk:
    3 Ampoul
  • Standards:
    USP reference standard
  • Description:
    Cumene
  • Pk Info:
    1.2 mL/ampule; 3 ampules
  • UN No.:
    1918
  • Cat. No.:
    1151709.
  • Description:
    Cumene
  • CAS No.:
    98-82-8

Specifications