Order Entry
Austria
Orders LinkContactUsLinkComponent
Acetone, Reference Standard

Acetone, Reference Standard

Acetone, USP reference standard, 1.5 mL/ampule; 3 ampules, 3
Catalog # 1006801.
Supplier:   USP
Acetone, Reference Standard
Catalog # 1006801.
Supplier:  USP
CAS Number:  
Acetone, Reference Standard
Acetone, Reference Standard

Acetone, USP reference standard, 1.5 mL/ampule; 3 ampules, 3

CAS Number:  
Price
437,00 €
Log in for availability
1 * 3 ST
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications

Specifications

  • Pk:
    3
  • Standards:
    USP reference standard
  • Description:
    Acetone
  • Pk Info:
    1.5 mL/ampule; 3 ampules
  • UN No.:
    1090
  • Cat. No.:
    1006801.
  • Formula:
    CH₃COCH₃
  • Description:
    Acetone
  • CAS No.:
    67-64-1

Specifications