Order Entry
Austria
Orders LinkContactUsLinkComponent
Acetazolamide, Reference Standards

Acetazolamide, Reference Standards

Supplier:   USP
Acetazolamide, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1005004.
  • 1005048.
  • 1005059.
  • Description:
  • Acetazolamide
  • Acetazolamide Related Compound D
  • Acetazolamide Related Compound E
  • Synonym(s):
  • 5-Amino-1,3,4-thiadiazole-2-sulfonamide
  • 5-Acetamido-1,3,4-thiadiazole-2-sulfonic acid potassium salt
  • CAS No.:
  • 59-66-5
  • 14949-00-9
  • Formula:
  • C₄H₆N₄O₃S₂

Specifications

Product Family Options

Product Information

  • Standards
    Pk
    Availability
    Price
  • 1005004.
    Acetazolamide
    Acetazolamide
    USP reference standard
    2 g
    Log in for availability
    Log in for availability
    1 * 2 g
    1 * 2 g - 412,00 €
    412,00 €
    1 * 2 g
       412,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1005004.
    Description
    Acetazolamide
    Synonym(s)
    CAS No.
    59-66-5
    Formula
    C₄H₆N₄O₃S₂
  • 1005048.
    Acetazolamide Related Compound D
    Acetazolamide Related Compound D
    USP reference standard
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 260,00 €
    1 260,00 €
    1 * 25 mg
       1 260,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1005048.
    Description
    Acetazolamide Related Compound D
    Synonym(s)
    5-Amino-1,3,4-thiadiazole-2-sulfonamide
    CAS No.
    14949-00-9
    Formula
  • 1005059.
    Acetazolamide Related Compound E
    Acetazolamide Related Compound E
    USP reference standard
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 360,00 €
    1 360,00 €
    1 * 25 mg
       1 360,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1005059.
    Description
    Acetazolamide Related Compound E
    Synonym(s)
    5-Acetamido-1,3,4-thiadiazole-2-sulfonic acid potassium salt
    CAS No.
    Formula
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.