Order Entry
Austria
Orders LinkContactUsLinkComponent
Acetaminophen, Reference Standards

Acetaminophen, Reference Standards

Supplier:   USP
Acetaminophen, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1003009.
  • 1003064.
  • 1003100.
  • Description:
  • Acetaminophen
  • Acetaminophen
  • Acetaminophen
  • Synonym(s):
  • p-nitrophenol
  • N-(4-chlorophenyl)acetamide
  • CAS No.:
  • 103-90-2
  • 100-02-7
  • 539-03-7
  • Formula:
  • C₈H₉NO₂
  • C₆H₅NO₃
  • C₈H₈ClNO
  • UN No.:
  • 1663

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1003009.
    Acetaminophen
    Acetaminophen
    USP reference standard
    Glass bottle for solids
    400 mg
    Log in for availability
    Log in for availability
    1 * 400 mg
    1 * 400 mg - 406,00 €
    406,00 €
    1 * 400 mg
       406,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1003009.
    Description
    Acetaminophen
    Synonym(s)
    CAS No.
    103-90-2
    Formula
    C₈H₉NO₂
    UN No.
  • 1003064.
    Acetaminophen
    Acetaminophen
    USP reference standard
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 150,00 €
    1 150,00 €
    1 * 50 mg
       1 150,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1003064.
    Description
    Acetaminophen
    Synonym(s)
    p-nitrophenol
    CAS No.
    100-02-7
    Formula
    C₆H₅NO₃
    UN No.
    1663
  • 1003100.
    Acetaminophen
    Acetaminophen
    USP reference standard
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 150,00 €
    1 150,00 €
    1 * 50 mg
       1 150,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1003100.
    Description
    Acetaminophen
    Synonym(s)
    N-(4-chlorophenyl)acetamide
    CAS No.
    539-03-7
    Formula
    C₈H₈ClNO
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.