Order Entry
Austria
Orders LinkContactUsLinkComponent
Acetaldehyde, Reference Standard

Acetaldehyde, Reference Standard

Acetaldehyde, USP reference standard, 3 x 1.1 mL, 3
Catalog # 1002811.
Supplier:   USP
Acetaldehyde, Reference Standard
Catalog # 1002811.
Supplier:  USP
CAS Number:  
Acetaldehyde, Reference Standard

Acetaldehyde, USP reference standard, 3 x 1.1 mL, 3

CAS Number:  
Price
392,00 €
Log in for availability
1 * 3 ST
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pk:
    3
  • Standards:
    USP reference standard
  • Description:
    Acetaldehyde
  • Pk Info:
    3 x 1.1 mL
  • UN No.:
    1089
  • Cat. No.:
    1002811.
  • Formula:
    C₂-H₄-O
  • Description:
    Acetaldehyde
  • CAS No.:
    75-07-0

Specifications