ContactUsLinkComponent
Clipped corner microscope slide, Snowcoat®
Clipped corner microscope slide, Snowcoat®
Catalog # LEIC3800381
Supplier:  LEICA MICROSYSTEMS
undefined
Clipped corner microscope slide, Snowcoat®
Catalog # LEIC3800381
Supplier:  LEICA MICROSYSTEMS
Specifications
  • Colour:
    White
  • Description:
    Snowcoat® clipped corner slide, colored ends
  • Pk:
    1.440
Specifications
About this item
Snowcoat® Clipped Corder Slides are clipped at 45° angles to help reduce glass breakage and fragmentation when loaded into slide printers.
  • Available in a variety of colours
  • Specialty painted surface
  • Efficient scanning
Specialty painted surface formulated to optimise Ink Jet printing and bar code scanning efficiency. These slides have been optimised for use with the Leica printer.