Order Entry
Austria
ContactUsLinkComponent
DSP (Dithiobis(succinimidyl propionate))
DSP (Dithiobis(succinimidyl propionate))
Catalog # BC07
Supplier:  G-Biosciences
CAS Number:  
undefined
DSP (Dithiobis(succinimidyl propionate))
Catalog # BC07
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Spacer Arm (Å):
    12
  • Reactive Toward:
    Primary Amines
  • Membrane Permeable:
    Yes
  • Water Soluble:
    No
  • Cleavable/ Reversible:
    Reducing Agents (Thiols)

Specification Test Results

About this item

 :
 :404.41 g/mol
CAS Number:57757-57-0
Formula:C₁₄H₁₆N₂O₈S₂
MDL Number:MFCD00042045