Order Entry
Austria
ContactUsLinkComponent
 
Ni-NTA His-Bind® Superflow™ Resin
 
Ni-NTA His-Bind® Superflow™ Resin

 

  • pH range:
    45 - 165 µm
  • Storage temperature:
    2 - 8 °C
  • Binding capacity:
    5 - 50 mg/mL

 

Frequently Bought Together