Order Entry
Austria
ContactUsLinkComponent
 
Desiccator cabinet, SICCO Tower Star
  BOHLV1938-07
 :  Bohlender
undefined
Desiccator cabinet, SICCO Tower Star
  BOHLV1938-07
 :  Bohlender

 

  • W×D×H ext.:
    310×375×735 mm
  • Description:
    Tower-Star desiccator for drying and storage (73 L)
  • Pk:
    1
  • Shelves:
    4
  • W×D×H int.:
    260×330×690 mm

 

 

Aluminium frame with panels made of acrylic glass, including four shelves made of polycarbonate, tray, hygrometer and desiccant, usable with up to 38 shelves, stackable.

  • Reinforced aluminium frame with acrylic panels
  • Door with magnetic catch and circular rubber seal
  • Easy to read electronic hygrometer
  • Variable adjustable shelves
  • Extractable shelves
  • Non-slip rubber feet
  • UVC 1

SICCO Desiccators are designed for storing or drying humidity sensitive products using silica gel. The tight fitting door protects the contents from contamination from the atmosphere. The controlled environment inside the desiccators is ideal for storing reference materials, retained samples, metallographic specimen, tobacco and DNA-samples.