ContactUsLinkComponent
Swinging bucket rotor assembly, SW 60 Ti, Optima
Swinging bucket rotor assembly, SW 60 Ti, Optima
Catalog # 521-3688
Supplier:  Beckman Coulter
undefined
Swinging bucket rotor assembly, SW 60 Ti, Optima
Catalog # 521-3688
Supplier:  Beckman Coulter
Specifications
  • Description:
    SW 60 Ti swinging bucket rotor assembly. Includes: SW 60 Ti rotor bucket set
  • Pk:
    1
  • For:
    Swinging bucket (ultracentrifugation)
  • Max. RPM:
    60000
  • No. of tubes accepted:
    8×5 ml
  • Radius:
    120,3
    mm