Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Chromatography Vial Closures
Caps for 13-425 screw thread vials
Caps for 13-425 screw thread vials
Caps for 13-425 screw thread vials
NSCAC4015-66A
:
THERMO NATIONAL SCIENTIFIC
Caps for 13-425 screw thread vials
NSCAC4015-66A
:
THERMO NATIONAL SCIENTIFIC
Cap Size:
13-425
Septum:
Red PTFE/White Silicone
Description:
Open top screw cap, phenolic cap type
Cap colour:
Black
Pk:
100