Rotor assembly, single lock, Avanti JA-25.50
521-3996
- Capacity:Nominal capacity
400 ml - Description:JA-25.50 fixed angle (high performance) aluminium rotor, with single lock lid, 8×50 ml, 25000 rpm, 75600 ×g. For use in Avanti J Series and J2 Series centrifuges
- Pk:1
- For:Major applications: Harvesting bacteria, processing tissue homogenates, subcellular particulates, routine pelleting such as precipitates and phase separations
- Max. RPM:25000
- No. of tubes accepted:8×50 ml/29×104 mm (8×50 ml/1×4")
- Radius:108 mm