Order Entry
Austria
ContactUsLinkComponent
Rotor assembly, Avanti JS-13.1
Rotor assembly, Avanti JS-13.1
Catalog # 521-3805
Supplier:  Beckman Coulter
undefined
Rotor assembly, Avanti JS-13.1
Catalog # 521-3805
Supplier:  Beckman Coulter

Specifications

  • Capacity:
    Nominal capacity
    300 ml
  • Description:
    JS-13.1 swinging bucket (high performance) aluminium rotor with carrying handle. For use in Avanti J Series and J2 Series centrifuges
  • Pk:
    1
  • For:
    Major applications: Harvesting bacteria, processing tissue homogenates, separating cell particulates. Isolation of cell membrane using density gradients
  • Max. RPM:
    13000
  • Radius:
    140 mm

Specifications

About this item