Stainless steel multi-bucket system
: CONTEC
Stainless steel cart with two buckets and downpress wringer.
- Includes two 27 L buckets and one cart with four non-marking, autoclavable casters
- Cart dimensions: 66×61×92 cm, with handle and casters
- Buckets, wringer, and cart are manufactured from 304 electropolished stainless steel and are fully autoclavable
- Buckets are deep drawn and seamless, reducing the risk of microbial contamination
- A lip around the top edge of the buckets allows for easy lifting
The stainless steel multi-bucket system offers maximum flexibility and can be used with two or three 27 L depending upon in-house protocols. An extendable frame pulls out of the front of the cart to accommodate an additional bucket. Lightweight and easy to maneuver, the cart is fully autoclavable and features non-marring wheels. The stainless steel downpress wringer is compatible with all sizes of VertiKlean mops and edgeless string mops.
Frequently Bought Together