Order Entry
Austria
ContactUsLinkComponent
 
Heating and cooling process thermostats, Integral XT
 :  LAUDA
New Product
 
undefined
Heating and cooling process thermostats, Integral XT
 :  LAUDA
New Product

 

  • Catalog No:
  • LAUDL002681
  • LAUDL002677
  • LAUDL002676
  • LAUDL002675
  • LAUDL002674
  • LAUDL002673
  • LAUDL002689
  • LAUDL002687
  • LAUDL002685
  • LAUDL002683
  • LAUDL002684
  • LAUDL002680
  • LAUDL002678
  • LAUDL002682
  • Model:
  • IN 2560 XTW
  • IN 750 XT
  • IN 550 XTW
  • IN 550 XT
  • IN 250 XTW
  • IN 150 XT
  • IN 1590 XTW
  • IN 590 XTW
  • IN 280 XTW
  • IN 8 XTW
  • IN 280 XT
  • IN 1850 XTW
  • IN 950 XTW
  • IN 4 XTW
  • Electrical:
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 230 V, 50 Hz
  • 230 V, 50 Hz
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 400 V, 3/PE 50 Hz and 460 V, 3/PE 60 Hz
  • 230 V, 50 Hz

 

Frequently Bought Together