Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Carbon disulphide ≥99.90%, SPECTRONORM® for spectroscopy
  85971.290
 :  
undefined
 
 
 
Carbon disulphide ≥99.90%, SPECTRONORM® for spectroscopy
  85971.290
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay :
    min. 99.90%
  • Water:
    0.01%
  • Evaporation residue:
    max. 0.0010%
  • Benzene (GC):
    max. 0.0030%
  • Toluene (GC):
    max. 0.0010%
  • Xylene (GC):
    max. 0.0010%
  • Alkalinity (meq/g):
    max. 0.0002
  • Acidity (meq/g):
    max. 0.0005
  • Color (APHA):
    max. 10
  • UV Transmission (382 nm):
    min. 10.00%
  • UV Transmission (388 nm):
    min. 35.00%
  • UV Transmission (403 nm):
    min. 80.00%
  • UV Transmission (410 nm):
    min. 90.00%
  • UV Transmission (500 nm):
    min. 99.00%

 

 

 :
 :76.14 g/mol
 :13,01821
 :46.2 °C (1013 hPa)
 :–111.6 °C
 :1.2632 g/cm³ (20 °C)
 :–30 °C
 :Ambient
 :200-843-6
 :1131
 :3 (6.1),I
 :Packaged under nitrogen.
 :GHS02
 :GHS08
 :GHS07
 :75-15-0
 :CS₂
 :MFCD00011321