ContactUsLinkComponent
Fixed-angle rotor package/assembly, TLA-100
Fixed-angle rotor package/assembly, TLA-100
Supplier:  Beckman Coulter
Certificates
undefined
Fixed-angle rotor package/assembly, TLA-100
Supplier:  Beckman Coulter
Specifications
  • Catalog No:
  • 521-3760
  • 521-3759
  • Capacity:
  • 4,0 ml
  • 4,0 ml
  • No. of tubes accepted:
  • 20×0 ml
  • 20×0 ml
  • Radius:
  • 38,9 mm
  • 38,9 mm
  • Max. RPM:
  • 100000
  • 100000