Zero pressure weather caps
Supplier: LABCONCO
Certificates
About this item
Weathercaps are constructed of strong, corrosion-resistant PVC.
Caps fits atop standard PVC duct, and permit vertical discharge of effluent air above roofline for dispersion away from the building. Caps add little static pressure to the exhaust system.
Frequently Bought Together