Order Entry
Austria
ContactUsLinkComponent
 
Heating and cooling process thermostats, Integral T
 :  LAUDA
 
undefined
Heating and cooling process thermostats, Integral T
 :  LAUDA

 

  • Catalog No:
  • LAUDL002670
  • LAUDL002664
  • LAUDL002663
  • LAUDL002669
  • LAUDL002668
  • LAUDL002667
  • LAUDL002666
  • LAUDL002665
  • Electrical:
  • 400 V, 3/PE, 50 Hz and 460 V, 3/PE, 60 Hz
  • 230 V, 50 Hz
  • 230 V, 50 Hz
  • 400 V, 3/PE, 50 Hz and 460 V, 3/PE, 60 Hz
  • 400 V, 3/PE, 50 Hz and 460 V, 3/PE, 60 Hz
  • 400 V, 3/PE, 50 Hz and 460 V, 3/PE, 60 Hz
  • 400 V, 3/PE, 50 Hz and 460 V, 3/PE, 60 Hz
  • 230 V, 50 Hz

 

Frequently Bought Together