ContactUsLinkComponent
Analytical 8-place titanium rotor, An-50 Ti
Analytical 8-place titanium rotor, An-50 Ti
Catalog # 521-4025
Supplier:  Beckman Coulter
undefined
Analytical 8-place titanium rotor, An-50 Ti
Catalog # 521-4025
Supplier:  Beckman Coulter
Specifications
  • Description:
    An-50 Ti analytical 8-place titanium rotor, requires XL-A/XL-I user interface
  • Environmentally Preferable:
  • Pk:
    1
  • For:
    Optima AUC and Optima AUC absorbance /interference centrifgue
  • Max. RPM:
    50000
  • No. of tubes accepted:
    8