Orders LinkContactUsLinkComponent
Inverted microscope platform, DMi8
Inverted microscope platform, DMi8
Leica DMi8
Catalog # 630-3468
Supplier:   LEICA MICROSYSTEMS
Inverted microscope platform, DMi8
Catalog # 630-3468
Supplier:  LEICA MICROSYSTEMS
Inverted microscope platform, DMi8
Inverted microscope platform, DMi8
Leica DMi8
Price
158 544,29 €
Log in for availability
1 * 1 SET
Product Details & Documents
Being ahead of the competition is what drives your business. No matter if you work in metallography, medical device manufacturing or microelectronics, speed is essential. Tailor this highly modular inverted microscope to your needs. Combine Leica optical quality, a wide range of contrast modes, and intuitive software in one system to help you speed up your workflow.
  • Speed up your workflow with inverted microscopy
  • Stay flexible with modular design
  • Easy reproducibility and documentation at a keystroke
  • See more details with unique UC-3D illumination
  • Individual functions support your workflow
From basic microscopy to high-end imaging – Leica DMi8 grows with ever-changing needs. Fully configurable with manual to motorized components, the DMi8 microscope allows you to build the imaging system for your research and budget needs from routine to live cell research. The inverted Leica DMi8 platform can be built up with a wide range of options covering all contrast methods and a comprehensive range of peripheral components. For maximum flexibility and upgradeability, choose the automated stand, which allows the addition of both manual and motorized components. When only limited capabilities are required, the coded or manual stands provide a solid core for basic applications.
Certifications: CE marked.
Ordering information: For more information and available components and options, contact your Avantor representative.
Specifications
  • Description:
    Leica DMi8
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Leica DMi8 T-House(0) for microscopes without fluorescence methods to hold all different transmitted light arms
    Leica DMi8 T-House(0) for microscopes without fluorescence methods to hold all different transmitted light arms
    LEIM11889036
    Log in for availability
    1 * 1 ST - 587,60 €