
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92921-24-9
:
Formula:C₁₆H₁₈N₂O₉S
MDL Number:MFCD00054978
:436.37 g/mol
Melting Pt:179 °C
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Maleimide, Sulfo-NHS Ester
- Spacer:Short
- Form:Powder
- Quantity:50 mg/1 g
- Type:Crosslinker
- Solubility:Water
Specification Test Results
Frequently Bought Together