Order Entry
Austria
ContactUsLinkComponent
Slc7a11 (NM_011990) mouse tagged ORF clone
Slc7a11 (NM_011990) mouse tagged ORF clone
Catalog # ORIGMG225346
Supplier:  OriGene
CAS Number:  
Slc7a11 (NM_011990) mouse tagged ORF clone
Catalog # ORIGMG225346
Supplier:  OriGene
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Description:
    Slc7a11 (NM_011990) mouse tagged ORF clone

Specifications

About this item

Slc7a11 (GFP-tagged) - Mouse solute carrier family 7 (cationic amino acid transporter y+ system) member 11 (Slc7a11), (10 µg).

  • Type: Mouse tagged ORF clone
  • Tag: TurboGFP
  • Symbol: Slc7a11
  • Synonyms: 9930009M05Rik; AI451155; sut; xCT
  • Vector: pCMV6-AC-GFP
  • E. coli Selection: Ampicillin (100 µg/ml)
  • Mammalian cell selection: Neomycin
  • Restriction sites: SgfI-MluI