Order Entry
Austria
ContactUsLinkComponent
Low pH SDS Sample Buffer
Low pH SDS Sample Buffer
Artikelnr. EKSIC44807
Supplier:  EKSIGENT
CAS Number:  
Low pH SDS Sample Buffer
Artikelnr. EKSIC44807
Supplier:  EKSIGENT
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Low pH SDS Sample Buffer
  • Pk Info:
    1× 140 ml
  • pH value:
    6,8

Specifications

About this item

SCIEX Low pH SDS Sample Buffer, 140 mL (100 mM Tris-HCl, pH 6.8, 1% SDS) allows you to use SCIEX’s Gold Standard CE-SDS with a low pH SDS sample buffer. Use of this buffer can increase non-reduced purity by decreasing method induced fragmentation. Less variability and lower RSDs means less troubleshooting and faster method development.