ContactUsLinkComponent
Vacuum pump systems, remote-controlled, type SC 820 G and SC 840 G, LABOPORT®
 :  KNF
undefined
Vacuum pump systems, remote-controlled, type SC 820 G and SC 840 G, LABOPORT®
 :  KNF
  • Catalog No:
  • 181-0648
  • 181-0649
  • Model:
  • SC 820 G
  • SC 840 G
  • Capacity:
  • Flow rate of up to 20 L/min
  • Flow rate of up to 34 L/min
  • Ultimate vacuum:
  • 6 mbar abs.
  • 6 mbar abs.
  • W×D×H:
  • 347×416×260 mm
  • 366×416×274 mm
Frequently Bought Together