Order Entry
Austria
ContactUsLinkComponent
Ammonium dihydrogen phosphate 100 g/L in nitric acid 1%, ARISTAR® matrix modifier solution, for graphite furnace AAS
Irritant
Ammonium dihydrogen phosphate 100 g/L in nitric acid 1%, ARISTAR® matrix modifier solution, for graphite furnace AAS
Catalog # 91275.154
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Ammonium dihydrogen phosphate 100 g/L in nitric acid 1%, ARISTAR® matrix modifier solution, for graphite furnace AAS
Catalog # 91275.154
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
ADR:8,III
 :115.03 g/mol
UN:1760
 :GHS07
Storage Temperature:Ambient
CAS Number:7722-76-1
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396