Drinking straws
Reusable drinking straws are made from borosilicate glass 3.3.
- Straight form, with fused edges
- Outer diameter: 9 mm
- Inner diameter: 6 mm
- Autoclavable
Ideal for use in hospitals, these straws are highly resistant against chemicals and thermal shock.
Frequently Bought Together