Orders LinkContactUsLinkComponent
ESD Safe rubber handle tweezers
ESD Safe rubber handle tweezers
Supplier:   OUTILS RUBIS
ESD Safe rubber handle tweezers
Supplier:  OUTILS RUBIS
Certificates
ESD Safe rubber handle tweezers
ESD Safe rubber handle tweezers
9 Options Available Below
Product Details & Documents
Streamlined tweezers with rubber handles to grant a secure grip, even after several hours of work.
  • Made from anti-magnetic stainless steel, with TPV rubber handles
  • Excellent ESD properties
  • Special streamlined shape fits the human hand perfectly
TPV rubber material has been developed with high-tech materials to provide heat insulating, effortless handling, extended durability, and ESD safe material (10⁷ Ohm). Rubies streamlined rubber handles are particularly recommended for cleanrooms and other controlled environments. They also provide good thermic insulation from heat, which makes them perfect for soldering and desoldering applications.
Product Family Options
Product Information
  • Type
    Length
    Tip width
    Thickness
    Availability
    Price
  • 232-0239
    Angled, bent, pointed ultra-fine tips
    Angled, bent, pointed ultra-fine tips
    5AR
    115 mm
    0,3 mm
    0,15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,00 €
    64,00 €
    1 * 1 ST
       64,00 €
  • 232-0238
    Angled, pointed ultra-fine tips
    Angled, pointed ultra-fine tips
    5A
    120 mm
    0,2 mm
    0,15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,40 €
    59,40 €
    1 * 1 ST
       59,40 €
  • 232-0236
    Straight, pointed ultra-fine tips
    Straight, pointed ultra-fine tips
    3C
    110 mm
    0,3 mm
    0,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,60 €
    53,60 €
    1 * 1 ST
       53,60 €
  • 232-0235
    Straight, pointed ultra-fine tips
    Straight, pointed ultra-fine tips
    3
    125 mm
    0,3 mm
    0,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,60 €
    53,60 €
    1 * 1 ST
       53,60 €
  • 232-0237
    Straight, needle pointed ultra-fine tips
    Straight, needle pointed ultra-fine tips
    5
    120 mm
    0,3 mm
    0,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,40 €
    59,40 €
    1 * 1 ST
       59,40 €
  • 232-0240
    Curved, pointed ultra-fine tips
    Curved, pointed ultra-fine tips
    7
    125 mm
    0,3 mm
    0,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,80 €
    62,80 €
    1 * 1 ST
       62,80 €
  • 232-0233
    Straight, round fine tips
    Straight, round fine tips
    2A
    125 mm
    2,0 mm
    0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,00 €
    53,00 €
    1 * 1 ST
       53,00 €
  • 232-0234
    Curved, round fine tips
    Curved, round fine tips
    2B
    125 mm
    2,0 mm
    0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,50 €
    61,50 €
    1 * 1 ST
       61,50 €
  • 232-0232
    Straight, pointed thick tips
    Straight, pointed thick tips
    00
    120 mm
    0,4 mm
    0,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,10 €
    55,10 €
    1 * 1 ST
       55,10 €