Ball / flange connections, DURAN®
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
Made from DURAN® tubing.
- High temperature resistance
- Very good chemical resistance
Manufactured to inch dimensions in accordance with DIN 12244 and DIN 12264.
Frequently Bought Together