ContactUsLinkComponent
Beaker tongs, stainless steel
Catalog # RSGA80.043.330
Supplier:  RSG
undefined
Beaker tongs, stainless steel
Catalog # RSGA80.043.330
Supplier:  RSG
Specifications
  • Length:
    300 mm
  • Pk:
    1
  • Spanning area:
    60 - 150 mm
Specifications
About this item
Beaker/cup tongs are made from stainless steel 1.4301.
  • Electrolytically polished
  • Jaws wrapped with organic fleece