ContactUsLinkComponent
AseptiQuik® G Connectors
AseptiQuik® G Connectors
Supplier:  CPC COLDER
Certificates
undefined
AseptiQuik® G Connectors
Supplier:  CPC COLDER
Specifications
  • Catalog No:
  • 212-0819
  • 212-0821
  • 212-0820
  • 212-0822
  • Max. operating pressure:
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Operating temperature:
  • 1...40 °C
  • 1...40 °C
  • 1...40 °C
  • 1...40 °C
Specifications
Connector cuffs
Product Information