ContactUsLinkComponent
MicroCNX™ connectors
MicroCNX™ connectors
Supplier:  CPC COLDER
Certificates
undefined
MicroCNX™ connectors
Supplier:  CPC COLDER
Specifications
  • Catalog No:
  • 212-0792
  • 212-0791
  • 212-0790
  • Max. operating pressure:
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Up to 4,1 bar; Up to 5,1 bar for 48 hours
  • Max. temperature:
  • 1...40 °C
  • 1...40 °C
  • 1...40 °C