MPX series ¹/₂" flow, quick disconnect coupling for bio and pharma
Lieferant: CPC COLDER
Zertifikate
Über diesen Artikel
These ¹/₂" flow quick disconnect couplings are manufactured specifically for the bio and pharma markets.
Materials of construction include animal free (BSE/TSE) USP class 6 polysulphone and polycarbonate, Buna - n and platinum cured USP class 6 silicone oring seals.
USP Class 6 and animal free (BSE/TSE) materials are appropriate for the Bio, Pharma and Life Sciences with single use dispoability in mind.
Certifications: USP Class VI, BSE/TSE free materials (ADCF)
Ordering information: Accessories not included
Frequently Bought Together