HFC39 series plastic couplings
These high flow couplings feature automatic shut-off valves which stop flow and eliminate the need for pinch clamps, stainless steel connectors or fittings.
They allow rapid disconnect and reconnect without the need for tools. Valved couplings stop fluid flow at the point of connection, reducing spills and related safety issues. Couplings feature an audible click when engaged, ensuring the user of a secure connection. Constructed of medical grade polysulphone (amber tint) with clear silicone seals. Couplings are sterilisable, reusable and compatible with virtually all existing processes.
: All products meet Class VI criteria and feature BSE-/TSE-free materials (ADCF).
- Catalog No:
- 212-0793
- 212-0797
- 212-0798
- 212-0800
- 212-0799
- 212-0794
- 212-0796
- 212-0795
- Max. operating pressure:
- Vacuum...8,62 bar
- Vacuum...8,62 bar
- Vacuum...8,62 bar
- Vacuum...8,62 bar
- Vacuum...8,62 bar
- Vacuum...8,62 bar
- Vacuum...8,62 bar
- Vacuum...8,62 bar
- Operating temperature:
- –40...+137,8 °C
- –40...+137,8 °C
- –40...+137,8 °C
- –40...+137,8 °C
- –40...+137,8 °C
- –40...+137,8 °C
- –40...+137,8 °C
- –40...+137,8 °C
Frequently Bought Together