Order Entry
Austria
ContactUsLinkComponent
Bromothymol blue solution R3 Reag. Ph. Eur.
Bromothymol blue solution R3 Reag. Ph. Eur.
Catalog # 84118.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bromothymol blue solution R3 Reag. Ph. Eur.
Catalog # 84118.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :624.39 g/mol
CAS Number:76-59-5
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872